Learn More
Wheaton™ Polypropylene White screw Cap
Product Code. 10332302 Shop All Wheaton Science Products Products
Click to view available options
Color:
White
Unit Size:
Pack of 1000
Product Code. 10332302
Brand: Wheaton™ 240805
Only null left
Specifications
Material (Liner) Metal Foil
Color White
Material Polypropylene
For Use With (Application) Screw caps for WHEATON liquid scintillation vials
Type Screw Cap
Closure Size 24-400