Learn More
VITLAB™ Polypropylene Test Tube Racks
Product Code. 10768514
Click to view available options
Positions:
32
Unit Size:
Pack of 5
Product Code. 10768514
Brand: VITLAB™ 3194948
Only null left
Specifications
Product Type Test Tube Rack
Color Blue
Holds Tubes (25 mm)
Material Polypropylene
Positions 32
Diameter (Metric) Opening 25 mm