Learn More
Ismatec™ Standard Tubing Connectors, Plastic
Click to view available options
Tubing Size:
1.5mm O.D.
2.5mm O.D.
3.0mm O.D.
4.0mm O.D.
5.0mm O.D.
6.0mm O.D.
8.0mm I.D.
Shape:
Straight
T-shape
Y-shape
Unit Size:
Pack of 10
Description
- Standard, non-reducing.
- Designed for Ismatec™ pump tubing.
- Available in different sizes, shapes, and compositions.
Specifications
Specifications
| Material | Acetal Resin (POM, Polyoxymethylene) |
| For Use With | For 3.0mm Tubing I.D. |
| Packaging Quantity | 10/Pk. |
| Tubing Size | 3.0mm O.D. |
| Shape | T-shape |
| Product Type | Standard Tubing Connector |