missing translation for 'onlineSavingsMsg'
Learn More

VITLAB™ Polypropylene Standard Joint Funnels

Product Code. 13365201
Click to view available options
Diameter (Metric)Top:
40mm
50mm
75mm
Length (Metric) Stem:
17mm
23mm
30mm
Description:
Thread size: NS 14/23
Thread size: NS 19/26
Thread size: NS 29/32
Height (Metric):
75 mm
95 mm
135 mm
Unit Size:
Each
Pack of 10
Pack of 5
This item is not returnable. View return policy

Product Code. 13365201

Brand: VITLAB™ 70694

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Fill multi-neck reaction flasks with standard neck joints. VITLAB™ Polypropylene Standard Joint Funnels are transparent, laterally flattened funnels that fit snugly into standard neck joints for fast, easy filling. Available for a variety of standard neck sizes.

Suitable for the filling of liquid or powdered reagents into a reaction flask during a reaction
  • Laterally flattened for use with multi-neck flasks
  • Suitable for standard joint necks of various sizes
  • Transparent material for easy monitoring of filling progress
  • Constructed of durable polypropylene
    • Chemically resistant for use with liquid or powdered reagents
    • Temperature resistant and autoclavable
    • FDA-grade quality

Specifications

Diameter (Metric)Top 75mm
Material Polypropylene
Height (Metric) 135 mm
Color Transparent
Description Thread size: NS 29/32
For Use With (Application) Suitable for the Filling of Liquid or Powdered Reagents Into a Reaction Flask During a Reaction
For Use With (Equipment) Multi-Neck Flasks and Single-Neck Flasks with Standard Joint Necks
Length (Metric) Stem 30mm
Quantity 5Pack
Product Type Standard Joint Funnel
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.