missing translation for 'onlineSavingsMsg'
Learn More

VITLAB™ Polypropylene Standard Joint Funnels

Product Code. 13355201
Click to view available options
Diameter (Metric)Top:
40mm
50mm
75mm
Length (Metric) Stem:
17mm
23mm
30mm
Description:
Thread size: NS 14/23
Thread size: NS 19/26
Thread size: NS 29/32
Height (Metric):
75 mm
95 mm
135 mm
Unit Size:
Each
Pack of 10
Pack of 5
This item is not returnable. View return policy

Product Code. 13355201

Brand: VITLAB™ 70594

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Fill multi-neck reaction flasks with standard neck joints. VITLAB™ Polypropylene Standard Joint Funnels are transparent, laterally flattened funnels that fit snugly into standard neck joints for fast, easy filling. Available for a variety of standard neck sizes.

Suitable for the filling of liquid or powdered reagents into a reaction flask during a reaction
  • Laterally flattened for use with multi-neck flasks
  • Suitable for standard joint necks of various sizes
  • Transparent material for easy monitoring of filling progress
  • Constructed of durable polypropylene
    • Chemically resistant for use with liquid or powdered reagents
    • Temperature resistant and autoclavable
    • FDA-grade quality

Especificaciones

Diameter (Metric)Top 50mm
Material Polypropylene
Height (Metric) 95 mm
Color Transparent
Description Thread size: NS 19/26
For Use With (Application) Suitable for the Filling of Liquid or Powdered Reagents Into a Reaction Flask During a Reaction
For Use With (Equipment) Multi-Neck Flasks and Single-Neck Flasks with Standard Joint Necks
Length (Metric) Stem 23mm
Quantity 10Pack
Product Type Standard Joint Funnel
Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado