missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VITLAB™ Polypropylene Standard Joint Funnels
Click to view available options
Diameter (Metric)Top:
40mm
50mm
75mm
Length (Metric) Stem:
17mm
23mm
30mm
Description:
Thread size: NS 14/23
Thread size: NS 19/26
Thread size: NS 29/32
Height (Metric):
75 mm
95 mm
135 mm
Unit Size:
Each
Pack of 10
Pack of 5
Description
Suitable for the filling of liquid or powdered reagents into a reaction flask during a reaction
- Laterally flattened for use with multi-neck flasks
- Suitable for standard joint necks of various sizes
- Transparent material for easy monitoring of filling progress
- Constructed of durable polypropylene
- Chemically resistant for use with liquid or powdered reagents
- Temperature resistant and autoclavable
- FDA-grade quality
Specifications
Specifications
| Diameter (Metric)Top | 75mm |
| Material | Polypropylene |
| Height (Metric) | 135 mm |
| Color | Transparent |
| Description | Thread size: NS 29/32 |
| For Use With (Application) | Suitable for the Filling of Liquid or Powdered Reagents Into a Reaction Flask During a Reaction |
| For Use With (Equipment) | Multi-Neck Flasks and Single-Neck Flasks with Standard Joint Necks |
| Length (Metric) Stem | 30mm |
| Quantity | 5Pack |
| Product Type | Standard Joint Funnel |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction