Learn More
Alpha Packaging™ Polypropylene Screw Caps
Product Code. 10044734
Click to view available options
Certifications/Compliance:
DIN 32
DIN 40
DIN 45
DIN 55
Use Alpha Packaging™ Polypropylene Screw Caps atop bottles containing analytical lab samples, especially those containing high purity chemicals, strong acids and solvents.
  • Autoclavable
  • Polypropylene
  • For use with screw-top bottles
Specifications
Material Polypropylene
No. per Pack 1 Each
Quantity 1 Each
Certifications/Compliance DIN 55
Product Type Screw Cap
Closure Material Polypropylene
Closure Type Screw Cap