missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Masterflex™ Concentric Reducer Fittings, Sanitary Clamp, Straight
Maintain sanitary conditions in your processes while decreasing diameter.
Brand: Masterflex™ 31805-06
Description
- Tri-Clamp™ concentric reducers decrease in diameter towards the center thus forming a conical cylinder
- Though both materials may be autoclaved, PVDF tolerates high temperatures better than PP does and will perform more consistently after repeated autoclaving
- Clamps and gaskets required for a complete system are sold separately
Specifications
Sanitary Fitting | |
Clamp Size: 1.5 x 0.75 in. | |
1 Pk. |
Polypropylene | |
-9°C | |
82°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Masterflex™ Concentric Reducer Fittings, Sanitary Clamp, Straight > Clamp Size: 1.5 x 0.75 in.
Spot an opportunity for improvement?Share a Content Correction