missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Witeg™ Polypropylene Pouring Ring
Click to view available options
Color:
Red
Description
- For GL45 Thread
- Polypropylene
- Various Colors
- Autoclavable
Specifications
Specifications
Color | Red |
Material | Polypropylene |
No. per Pack | 10Pack |
Quantity | 10Pack |
Autoclavable | Autoclavable |
Product Type | Pouring Ring |
For Use With (Equipment) | For GL 45 thread |
Closure Material | Polypropylene |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction