Learn More
BrandTech™ VITLAB™ Polypropylene Pipet Fillers
Designed to fit all glass and plastic pipets. VITLab™ Polypropylene Pipet Fillers are color coded for easy volume identification.
Brand:  BrandTech™ 002524
Encompass
 View more versions of this product
Product Code. 10662631
  • € 20.61 / Each
Estimated Shipment: 10-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For pipetting liquids
  • Slow rotation of the actuator-wheels draws liquid into the pipette
  • Pressing the air bleed valve automatically empties the pipette without returning the piston
Specifications
Specifications
Polypropylene
1 Each
Pipette Filler
Pipetting liquids
Red
Pipettes, 25 mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers