Learn More
Brady™ Polypropylene: ISO Safety Sign - Door opens by pushing on the right-hand side
Sign compliant to ISO 7010.
Brand: Brady™ 831246
This item is not returnable.
View return policy
Description
- Polypropylene
- For indoor and outdoor use
Specifications
ISO 7010 | |
Polypropylene | |
Indoor and Outdoor | |
1.501 mm | |
-40°C to 120°C | |
Global |
PIC E023-200x200-PP-CRD/1 | |
1 Ea. | |
200 mm | |
200 mm | |
Square | |
White on Green |