Learn More
Buerkle™ Polypropylene Straight Connectors with Conical Nozzles
Shop All Bürkle Products
Click to view available options
For Use With:
11 to 14 mm Hose
13 to 15 mm Hose
3 to 5 mm Hose
5 to 7 mm Hose
7 to 10 mm Hose
9 to 12 mm Hose
Unit Size:
Pack of 10
Description
Cylindrical straight hose connector with conical nozzles
- Polypropylene
- Autoclavable to 121°C
- Suitable for foodstuff
- Available in a wide range of diameters for varying hose sizes
Specifications
Specifications
| Certifications/Compliance | EU Foodstuffs, RoHS |
| Autoclavable | Autoclavable |
| Material | Polypropylene |
| For Use With | 7 to 10 mm Hose |
| Packaging Quantity | 10/Pk.; 10/Scale |
| Shape | Straight |
| Color | White |
| Product Type | Hose Tubing Connector |