Learn More
Brady™ Polypropylene Gate Valve Lockout
Click to view available options
Color:
Blue
Green
Red
Yellow
Width (Metric):
120.1 mm
184.8 mm
224.1 mm
318.3 mm
405.1 mm
Height (Metric):
80 mm
141.7 mm
181 mm
276.2 mm
363.5 mm
Unit Size:
Each
Description
- Our gate valve lockouts have an unique knockout centre accommodates OS and Y or ''rising stem'' valves.
- Lockouts accept locks with shackles up to 9.53 mm diameter.
- Danger labels in English, Spanish and French are included.
Specifications
Specifications
| Product Type | Gate Valve Lockout |
| Alternate Part Number | GATE VALVE LOCKOUTS GVLO 10-13 RED |
| Color | Red |
| Material | Polypropylene |
| Certifications/Compliance | OSHA 1910.147 |
| Description | Brady Gate Valve Lockouts |
| Diameter (English) | 13.96 in |
| Diameter (English) Shackle | 0.38 in. |
| Diameter (Metric) | 354.58 mm |
| Diameter (Metric) Shackle | 9.7 mm |
| Show More |