Learn More
BRAND™ Polypropylene Funnel with Fluted Interior
Product Code. 10433711 Shop All Brand Products
Click to view available options
Diameter (Metric)Top:
65 mm
120 mm
150 mm
Diameter (Metric) Stem:
9 mm
11 mm
14 mm
Unit Size:
Each
Pack of 20
Product Code. 10433711
Brand: BRAND™ 148237
Only null left
Add to basket
Add to basket
Polypropylene construction. Short wide stem provides smooth, quick funneling.
  • Polypropylene construction
  • Short, wide stem
  • Includes internal fluting
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric)Top 120 mm
Diameter (Metric) Stem 11 mm
Material Polypropylene
Length (Metric) Stem 85 mm