Learn More
Carl Roth™ Polypropylene Disposal Bags
Brand:  Carl Roth™ H736.1
Encompass_Preferred
 View more versions of this product
Product Code. 10030672
  • € 50.90 / Pack of 500
Estimated Shipment: 10-02-2025
to see stock.
Explore more special offers
Specifications
Waste Disposal Bag
Ideal for laboratory waste such as used pipettor tips, test tubes and vials.
30 μm
300 mm
Single-use
Polypropylene
300 x 200mm
200 mm