Learn More
Buerkle™ Closing Cap with Inner Thread
Shop All Bürkle Products
Click to view available options
Thread Size:
1/2 in.
3/4 in.
Unit Size:
Each
Description
- Polypropylene construction
- Autoclavable to 121°C
- Safe for use with foodstuffs
- Supplied in packs of 10
Specifications
Specifications
| Certifications/Compliance | EU Foodstuffs |
| Autoclavable | Autoclavable |
| Material | Polypropylene |
| Thread Size | 1/2 in. |
| Product Type | Closing Cap |