Learn More
Macherey-Nagel™ Polypropylene Chromabond™ C8 SPE Column
Shop All Macherey-Nagel Products
Click to view available options
Bed Weight:
100 mg
200 mg
500 mg
Column Capacity (Metric):
1 mL
3 mL
6 mL
Unit Size:
Each
Pack of 100
Pack of 50
Specifications
Specifications
| Base Material | Silica |
| Carbon Load | 8% |
| Column Capacity (Metric) | 6 mL |
| Column Type | SPE |
| Bed Weight | 500 mg |
| For Use With (Application) | Pesticides, PCBs Applications |
| Particle Size | 45 μm |
| Type | Solid Phase Extraction Column |
| Endcapped | No |
| Pore Size | 60 μm |
| Show More |