Learn More
BRAND™ Polypropylene Burette Clamps
Shop All Brand Products
Click to view available options
For Use With (Equipment):
For use with 1 Burette
For use with 2 Burettes
Unit Size:
Each
Description
- Polypropylene construction
- White
- For use with support rods 8 to 14mm dia.
Specifications
Specifications
| Color | White |
| For Use With (Equipment) | For use with 1 Burette |
| Material | Polypropylene |
| No. per Pack | 1 Each |
| Type | Buret Holder |