Learn More
Poulten & Graf GmbH Polypropylene Adaptor for FORTUNA™ Dispenser with Thread
Complete a FORTUNA™ Dispenser with an adapter that comes in a wide selection of threads. Poulten & Graf GmbH Polypropylene Adaptor for FORTUNA™ Dispenser with Thread enables smaller containers to be supported on top the existing reservior.
Brand:  Poulten & Graf GmbH 101 09030
 View more versions of this product
Product Code. 10024750
  • € 13.10 / Each
Estimated Shipment: 24-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Suitable for OPTIFIX and POLYFIX dispenser
  • Made of Polypropylene
  • Adapter with threads
  • Can be used for most chemicals and liquids
Specifications
Specifications
Adapters for OPTIFIX and POLYFIX Dispensers
White
Polypropylene
1
Autoclavable
Adapters for OPTIFIX and POLYFIX dispensers
OPTIFIX and POLYFIX dispensers
GL 30
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers