Learn More
Eppendorf™ Polypropylene Adaptor
Product Code. p-8004484 Shop All Eppendorf Products
Click to view available options
For Use With (Equipment):
Bottle Thread 32 to 28 mm
Bottle thread 25 mm
Bottle thread GL 32 to GL 27
Bottle thread GL 32 to GL 45
Bottle thread GL 45 to GL 35
Bottle thread GL 45 to GL 38
Bottle thread GL 45 to S 40
Bottleneck 19/26
Thread GL 32 to NS 24/29
Thread GL 32 to NS 29/32
Unit Size:
Each
Product Code. 10023211
Brand: Eppendorf™ 4960800040
Only null left
Specifications
Product Type Adapter
Material Polypropylene
For Use With (Equipment) Bottle thread 25 mm
Quantity 1 Ea.