Learn More
Kinematica™ Polymix™ PX-MFC 90 D Hammer or Cutting Mill
Shop All Kinematica Products
Click to view available options
Product Type:
Blade Grinding Mill
Hammer Grinding Mill
Plug Type:
EU plug
missing translation for 'unitSize'
Each
Description
Polymix™ MFC 90 D features easy to change grinding attachments and sieves extending the range of samples that can be processed. Simple handling, high user safety and efficient grinding results are just a few of the advantages of this mill.
- Working volume approx. 300mL (volume of the funnel)
- 600W high frequency 3 phase motor with speed regulation from 50rpm up to 6, 000rpm
- Integrated overheating, overload and blocking protection
- Safety switch for automatic off
- Energy saving through automatic standby mode
- Hammer grinding or blade grinding attachment
- Sieve size 2mm.
Specifications
Specifications
| Product Type | Hammer Grinding Mill |
| Dimensions (L x W x H) | 325 x 251 x 480 mm |
| Electrical Requirements | 230 V/GB, 50 Hz, 1000 W Motor Input Power |
| For Use With (Application) | Dry sample grinding |
| Hertz | 50/60 Hz |
| Plug Type | EU plug |
| Voltage | 230 V |
| Watts | 1000 W |
| Weight (Metric) | 12 kg |