Learn More
Macherey-Nagel™ Polygram POLYAMIDE-6 Polyester Sheets
Shop All Macherey-Nagel Products
Click to view available options
Dimensions (L x W):
5 x 20 cm
20 x 20 cm
Unit Size:
Pack of 25
Pack of 50
Specifications
Specifications
| Quantity | 50 Pk. |
| Type | Polyester Sheet |
| Thickness (Metric) | 0.1 mm |
| Dimensions (L x W) | 5 x 20 cm |