Learn More
Scancell Folien Vertriebs™ Polyethylene Ziplock Bag
Click to view available options
Dimensions (L x W):
150 x 220 x 0.05mm
400 x 500 x 0.05mm
Unit Size:
Pack of 100
Pack of 1000
Specifications
Specifications
| Color | Clear |
| Material | Polyethylene |
| Thickness (Metric) | 0.050 mm |
| Quantity | 100 Pack |
| Dimensions (L x W) | 150 x 220 x 0.05mm |
| Product Type | Ziplock Bag |