Learn More
Ratiolab™ Polyethylene Scintillation Vial
Product Code. 10359970
Click to view available options
Color:
Natural
Unit Size:
Pack of 1000
Product Code. 10359970
Brand: Ratiolab™ 5810100
Only null left
Specifications
Color Natural
Diameter (Metric) Outer 27 mm
Material Polyethylene
Volume (Metric) 20 mL
For Use With (Application) For use with robotic analytical systems
No. per Pack 1000
Closure Type Leakproof Cap
Height (Metric) 60 mm
Quantity 1000 Pk.
Type Scintillation Vial