Learn More
Masterflex™ Polyethylene Male Pipe Adapter, pack of 100
Designed for quick and easy assembly without tools
Brand:  Masterflex™ 30625-54
 View more versions of this product
Product Code. 11790109
  • € 120.00 / Pack of 100
Estimated Shipment: 28-07-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Polyethylene Male Pipe Adapter
  • Adapts from a NPT male thread to a hose barb
  • Barbed fittings are designed for quick and easy assembly without tools
  • Combine these simple, economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30psi)
  • Available in packs of ten
Specifications
Specifications
1/4 in. NPT(M) x 1/2 in. ID Tubing
HDPE
1/2 in.
1/4 in. NPT(M)
Ethylene Oxide
Straight
Male Pipe Adapter
Not Autoclavable
100 Fittings
1/2 in.
1/2 in.
1/4 in. NPT(M)
Translucent
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Masterflex™ Polyethylene Male Pipe Adapter, pack of 100 > 1/4 in. NPT(M) x 1/2 in. ID Tubing