Learn More
Buerkle™ Polyethylene Flat Seal
Product Code. 10458994 Shop All Bürkle Products
Click to view available options
Model:
1 in. Thread
3/4 in. Thread
Unit Size:
Pack of 10
Product Code. 10458994
Brand: Buerkle™ 85380034
Only null left
Add to basket
Add to basket
Flat seals made of Polyethylene, transparent fitting outer or inner thread.
  • PE
TRUSTED_SUSTAINABILITY
Specifications
Color Transparent
Description 3/4 in. thread
Model 3/4 in. Thread
Material Polyethylene
Type Seal, Flat