Learn More
Kartell™ Ranvier Type Dropping Bottle
Click to view available options
Capacity (Metric):
50 mL
100 mL
Unit Size:
Pack of 10
Description
- Comes with PE tube and rubber teat
- Suitable for foodstuff
Specifications
Specifications
| Capacity (Metric) | 100 mL |
| Color | Neutral |
| For Use With (Application) | Foodstuff |
| Includes | Polyethylene tube, Rubber teat |
| Material | Polyethylene |
| Quantity | 10Pack |
| Product Line | Bottle |
| Height (Metric) | 115 mm |
| Product Type | Dropping Bottle |
| Diameter (Metric) | 43 mm |