Learn More
Kartell™ Polyethylene Disc Membrane
These discs are made of polyethylene and have good chemical resistance. Kartell™ Polyethylene Disc Membrane is used to separate the filter paper from the funnel plate when working with a Buchner Funnel.
Brand: Kartell™ 0083800
9.79 EUR valid until 2025-03-30
Use promo code "24134" to get your promotional price.
This item is not returnable.
View return policy
Description
- Useful interface between filter paper and perforated Buchner plate
- Avoids clogging of filter holes
Specifications
Screen Filter | |
Polyethylene | |
Interface between filter paper and perforated Buchner plate | |
Circle |
Disc | |
300 μm | |
10/Pack | |
80 mm |
Product Content Correction
Product Title
Kartell™ Polyethylene Disc Membrane > Diameter: 80 mm