missing translation for 'onlineSavingsMsg'
Learn More

Shield Medicare™ Klerwipe™ Sterile Dry Wipes

For use in cleaning and disinfection procedures. Shield Medicare Klerwipe™ Sterile Dry Wipes are highly absorbent, making them useful both for applying cleaning solutions and disinfectants and to clean up spills.

Brand:  Shield Medicare™ 3035500

Product Code. 15227704

  • € 101.00 / Pack of 10
Estimated Shipment: 14-10-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Packed and double bagged in Grade C (ISO Class 7) cleanroom before gamma irradiation at 25kGy
  • 115gsm circular knit polyester with laser sealed edges
  • Made from 68gsm binder free, hydroentangled cellulose/polyester blend
  • Made from non-abrasive polyurethane foam
  • Suitable for cleanroom use
  • Flexible Application
  • Suitable for direct product contact use
Specifications

Specifications

Dry Wipe
200 mm
Low lint
230 x 230 mm
100 Pk.
200 mm
Polyester
EN ISO 9001:2008, EN ISO 13485:2003
Suitable for cleanroom use
Sterile
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers