Learn More
Brady™ Polyester High Performance Tape
Product Code. 11599233
Click to view available options
Dimensions (LxW):
30 m x 13 mm
30 m x 29 mm
30 m x 57 mm
30.48 m x 12.7 mm
30.48 m x 28.58 mm
30.48 m x 57.15 mm
Color:
Black
Blue
Brown
Gray
Green
Light Blue
Ochre
Orange
Pink
Purple
Red
Sky Blue
Tan
Transparent
White
Yellow
Unit Size:
Each
Product Code. 11599233
Brand: Brady™ 076636
B-569 Polyester High Performance Tape adheres to smooth surfaces with the same durability as B-595.
  • B-569 adheres to smooth surfaces with the same durability as B-595.
Specifications
Certifications/Compliance Compliant with Directive 2011/65/EU and 2015/863
Description GlobalMark High Performance Polyester Tape
Adhesive Type Permanent Acrylic, Pressure-sensitive
Coating None
Color Orange
Finish Gloss
Format Continuous
Material Polyester
Printable Sides Single-side Printable
Print Technology Thermal Transfer
Product Type Label
Sterility Non-sterile
For Use With (Application) Facility Identification, Pipe and valve marking
For Use With (Equipment) GlobalMark Printer
Gloss 80 Gardner Units
Labels per Row 1
Min. Operating Temperature (Metric) -40°C
No. per Pack 1 Roll/Box
Quantity 1 Ea.
Temperature Range (English) -40°F to +212°F
Temperature Range (Metric) -40°C to +100°C
Thickness (English) 0.003 in.
Thickness (Metric) 0.076 mm
Dimensions (LxW) 30.48 m x 57.15 mm
Length (English) 100 ft.
Length (Metric) 30.48 m
Width (English) 2.25 in.
Width (Metric) 57.15 mm