Learn More
Brady™ Polyester High Performance Tape
Click to view available options
Dimensions (LxW):
30 m x 13 mm
30 m x 29 mm
30 m x 57 mm
30.48 m x 12.7 mm
30.48 m x 28.58 mm
30.48 m x 57.15 mm
Color:
Black
Blue
Brown
Gray
Green
Light Blue
Ochre
Orange
Pink
Purple
Red
Sky Blue
Tan
Transparent
White
Yellow
Unit Size:
Each
Description
- B-569 adheres to smooth surfaces with the same durability as B-595.
Specifications
Specifications
| Certifications/Compliance | Compliant with Directive 2011/65/EU and 2015/863 |
| Description | GlobalMark High Performance Polyester Tape |
| Adhesive Type | Permanent Acrylic, Pressure-sensitive |
| Coating | None |
| Color | Sky Blue |
| Finish | Gloss |
| Format | Continuous |
| Material | Polyester |
| Printable Sides | Single-side Printable |
| Print Technology | Thermal Transfer |
| Show More |