Learn More
Velp Scientifica™ Polycarbonate Tank
Velp Scientifica™ Polycarbonate Tank is a fundamental accessory of the Velp Scientifica™ (OCB) Open Circulating Bath.
€ 245.00 - € 268.00
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
15339564
|
Velp Scientifica™
A00000090 |
5 L |
€ 245.00
Each |
Estimated Shipment: 16-01-2025
Log in to see stock. |
|||||
15349564
|
Velp Scientifica™
A00000091 |
7 L |
€ 268.00
Each |
Estimated Shipment: 23-01-2025
Log in to see stock. |
|||||
Description
- Completely transparent for a constant visual control
Specifications
OCB | |
Tank | |
Polycarbonate |