Learn More
Masterflex™ Polycarbonate Luer Fittings
Control flow effectively with these luer fittings Cole-Parmer™ Polycarbonate Luer Fittings are available in variety of styles to meet your application's needs.
€ 9.90 - € 61.25
Specifications
Material Polycarbonate
Product Type Luer Fitting
View More Specs
Products 2
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
11783924
View Documents
Masterflex™
06464-95
male lock x male lock, rotating This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11710009
View Documents
Masterflex™
30600-50
female x male lock, rotating, Non-sterile This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
Both straight and elbow connectors are available. Hose barbs are available from 1/16 to ¼ in. with either male or female luer connections.
Choose from several different barb styles:
  • Standard hose barb for use with flexible tubing
  • Smooth bore hose barb to reduce flow turbulence and risk of line contamination
  • Wide-bore hose barb for enhanced fluid path flow
  • Low-profile hose barbs for use with semi-rigid tubing such as nylon, polyethylene and polypropylene
Specifications
Specifications
Polycarbonate
Luer Fitting