accessibility menu, dialog, popup
Test
Search
Order Status
Quick Order
Support
Sign In
chevron-down
chevron-down
Learn More
Home
Products
Specialty Lab Equipment, Instruments, and Apparatuses
Laboratory Spatulas
VITLAB™ Polyamide Double Spoons
15282779
Print
VITLAB™ Polyamide Double Spoons
Length: 150mm
Brand:
VITLAB™ 80594
View more versions of this product
Product Code.
15282779
€ 25.75
/ Pack of 10
Quantity
Estimated Shipment: 17-12-2024
Log in
to see stock.
Explore more special offers
Certificates
Special Offers
Specifications
Length (Metric)
150 mm