Learn More
Buerkle™ Polyacetal Hose Clips
Product Code. 10054083 Shop All Bürkle Products
Click to view available options
Tubing Size:
10 to 11 mm
11 to 12 mm
12 to 14 mm
13 to 15 mm
15 to 17 mm
17 to 19 mm
18 to 20 mm
20 to 22 mm
23 to 25 mm
6 to 6.5 mm
9 to 10 mm
Unit Size:
Pack of 10
Product Code. 10054083
Brand: Buerkle™ 86780010
Only null left
Add to basket
Add to basket
Use these Buerkle™ Polyacetal Hose Clips to clamp down on tubing for a sure and secure fit.
  • Hose clips made of Polyacetal (POM)
  • Featured in varying diameters
  • POM is white, sterilizable, does not rust, permanently elastic and hygienic.
TRUSTED_SUSTAINABILITY
Specifications
Material Polyacetal
For Use With Hoses
Packaging Quantity 10 Pc./Pk.
Tubing Size 9 to 10 mm
Color White
Product Type Hose Clip