missing translation for 'onlineSavingsMsg'
Learn More

Poly(propylene glycol), average M.W. 425, Thermo Scientific Chemicals

€ 22.80 - € 541.70

Chemical Identifiers

CAS 25322-69-4
Molecular Formula (C3H6O)n
Molecular Weight (g/mol) 134.18
MDL Number MFCD00084446
InChI Key DNIAPMSPPWPWGF-UHFFFAOYNA-N
Synonym polypropylene glycol, polyoxypropylene, 2-2-hydroxypropoxy propan-1-ol, emkapyl, lineartop e, niax ppg, polyoxypropylene glycol, methyloxirane homopolymer, desmophen 360c, 2-2-hydroxypropoxy-1-propanol
PubChem CID 32881
IUPAC Name 2-(2-hydroxypropoxy)propan-1-ol
SMILES [H]OC(C)CO
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10154400
View Documents
Thermo Scientific Acros
192150050
5 g Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10338190
View Documents
Thermo Scientific
192152500
250 g Glass bottle
€ 76.38
250g
Estimated Shipment: 27-01-2025
to see stock.
Add to basket
 
10479890
View Documents
Thermo Scientific Acros
192155000
500 g Glass bottle
€ 147.74
500g
In Stock
to see stock.
Add to basket
 
11916981
View Documents
Thermo Scientific Acros
192150025
2.5 kg Glass bottle
€ 541.70
2.50kg
In Stock
to see stock.
Add to basket
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

25322-69-4
134.18
DNIAPMSPPWPWGF-UHFFFAOYNA-N
32881
[H]OC(C)CO
(C3H6O)n
MFCD00084446
polypropylene glycol, polyoxypropylene, 2-2-hydroxypropoxy propan-1-ol, emkapyl, lineartop e, niax ppg, polyoxypropylene glycol, methyloxirane homopolymer, desmophen 360c, 2-2-hydroxypropoxy-1-propanol
2-(2-hydroxypropoxy)propan-1-ol

Specifications

25322-69-4
-25.0°C
H[OCH(CH3)CH2]nOH
1.0000g/mL
Solubility in water: completely soluble. Other solubilities: soluble in ethanol, acetone, benzene, toluene,, chloroform, dioxane, ether, thf
[H]OC(C)CO
134.18
185°C
1
Poly(propylene glycol), Average M.W. 425
200.0°C
(C3H6O)n
MFCD00084446
polypropylene glycol, polyoxypropylene, 2-2-hydroxypropoxy propan-1-ol, emkapyl, lineartop e, niax ppg, polyoxypropylene glycol, methyloxirane homopolymer, desmophen 360c, 2-2-hydroxypropoxy-1-propanol
DNIAPMSPPWPWGF-UHFFFAOYNA-N
2-(2-hydroxypropoxy)propan-1-ol
32881
1.4465 to 1.4505
80 mm2/s (25°C)
Videos
Safety and Handling

Safety and Handling

missing translation for 'rtecsNumber' : OE5784360

missing translation for 'tsca' : TSCA

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.