missing translation for 'onlineSavingsMsg'
Learn More

SCAT Politainer (Foldable Waste Container)

Product Code. 10579490
Click to view available options
Capacity (Metric):
5 L
10 L
20 L
Closure Size:
GL38
S60/61
Dimensions (L x W x H):
178 x 178 x 178 mm
228 x 228 x 228 mm
285 x 285 x 285 mm
This item is not returnable. View return policy
This item is not returnable. View return policy

The SCAT Politainer - also known as folding canisters - is a combination of a flexible plastic bag and a stable UN-approved outer carton. Storage in the unfilled state is possible in a very small space and after filling, safe storage or transport is guaranteed.

  • Consists of LDPE and EVA copolymer, which offers optimum protection against volatile solvents
  • Available with the frequently used threads GL38 and S60/61 and with volumes of 5, 10 and 20 L

SCAT Politainers are an ideal way of not having to dispense with safe storage even in limited spaces

Specifications

Capacity (English) 2.6 gal.
Capacity (Metric) 10 L
Closure Size GL38
Color White (Translucent)
Description Politainer 10 L, GL38, PE-LD/EVA copolymer, foldable, dimensions (W x H x D): 228 x 228 x 228 mm
Dimensions (L x W x H) 228 x 228 x 228 mm
For Use With (Application) Waste Disposal System
For Use With (Equipment) Cardboard
Includes UN-Y approval in combination with box
Material LDPE
Mouth Style GL38 Thread
Product Type Canister
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'