Learn More
Cole-Parmer™ PolarSafe™ Stainless Steel Liquid Nitrogen Refill Tank
Shop All Cole-Parmer Products
Click to view available options
Capacity (Metric):
30 L
50 L
Unit Size:
Each
Description
- Durable welded stainless steel construction ensures long-lasting performance and reduced evaporation loss
- Unique design utilizes pressure from LN2 to discharge LN2 to other containers
- Use as a self-pressurized LN2 backup system for cryogenic freezers
- 5 year vacuum warranty
Specifications
Specifications
| Capacity (Metric) | 30 L |
| Depth (English) | 23 in. |
| Depth (Metric) | 58.42 cm |
| Diameter (English) Inner | 17.9 in. |
| Diameter (English) Outer | 23 in. |
| Diameter (Metric) Inner | 45.466 cm |
| Diameter (Metric) Outer | 58.42 cm |
| Material | Stainless Steel |