Learn More
Cole-Parmer™ PolarSafe™ Dewar and Traceable™ Temperature Data Logger Bundles
Product Code. 16602874 Shop All Cole-Parmer Products
Click to view available options
Capacity (Metric):
2 L
30 L
35 L
47 L
95 L
115 L
Unit Size:
Each
Product Code. 16602874
Brand: Cole-Parmer™ 4426071
Confidently store and monitor temperature-sensitive samples
  • Improve sample protection by maintaining specific temperature ranges for extended time periods
  • Maintain sample integrity with Traceable™ temperature monitoring
  • Data logger is provided with a 2-year NIST-traceable calibration certificate
Specifications
Capacity (Metric) 95 L
Depth (English) 31 in.
Depth (Metric) 78.74 cm
Diameter (English) Inner 8.5 in.
Diameter (English) Outer 33.5 in.
Diameter (Metric) Inner 21.6 cm
Diameter (Metric) Outer 85.09 cm
Material Aluminum