Learn More
Schneider Gerd™ Pointed Tweezer
Product Code. 10585334
Click to view available options
Length (Metric):
100 mm
115 mm
Design:
Bent
Spring Steel
Finish:
Teflon coated
Unit Size:
Each
Product Code. 10585334
Brand: Schneider Gerd™ 165
Only null left
Add to basket
Add to basket
Use these forceps for a variety of applications. Schneider Gerd™ Pointed Tweezers are ergonomically designed for optimal grip and precision.
  • Pointed tweezers
  • Ideal for dissection and general lab applications
Specifications
Design Bent
Length (Metric) 100 mm
Product Type Tweezers, Pointed
Finish
For Use With (Application) For Laboratory use