Learn More
S Murray and Co™ Platinum Wire Holder
Use this wire holder for inoculation wire or platinum wire up to 0.56mm in diameter. S Murray™ Platinum Wire Holders are fully guaranteed in normal use and subject to stringent quality inspection procedures.
Brand:  S Murray and Co™ L112/03
Product Code. 12794406
  • € 32.50 / Pack of 5
Estimated Shipment: 20-05-2025
to see stock.
Explore more special offers
Add to basket
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Description
Description
  • 200mm
  • Pack of 10
Specifications
Specifications
Wire Holder
Aluminum Alloy
200 mm
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
S Murray and Co™ Platinum Wire Holder > Length: 200mm