Learn More
Platinum Iridium wire, 0.127mm (0.005in) dia, annealed, 99.9% (metals basis excluding precious metals)
Product Code. p-7058683
missing translation for 'orderingAttributeHoverText'
Quantity:
10 cm
50 cm
missing translation for 'unitSize'
10cm
50cm
Product Code. 11364658
missing translation for 'mfr': Thermo Scientific Alfa Aesar 043019.BS
Request Bulk or Custom Format
Vacuum evaporation deposition, scanning and tunneling microscopy, corrosion resistant electrodes.Platinum-iridium wire is used in platinum jewellery, particularly watch cases. They have been used for spinnerets in the manufacture of synthetic fibres, in metrology and alloy is fabrication of metal microelectrodes. It is used in electrical stimulation of nervous tissue and electrophysiological recordings. Pt/Ir alloy has been used in mechanical and electrochemical devices.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Vacuum evaporation deposition, scanning and tunneling microscopy, corrosion resistant electrodes
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Platinum Iridium wire
Name Note 0.127mm (0.005 in.) dia., Annealed, Temper: soft
Avg. Mol. Wt. or Mol. Wt. Range Pt:Ir; 90:10 wt%
Quantity 10 cm
Odor Odorless
Weight ≈0.03g/10cm
Form Wire
Assay Percent Notes (metals basis excluding precious metals)
MDL Number MFCD02091683
Solubility Information Insoluble in water and acid.
Percent Purity 99.9%