Learn More
Kartell™ Plastilab™ Untapered T-Shaped Tubing Connectors
BIOPHARMA
Designed with serrations to hold hoses tightly and prevent slipping.
Brand:  Kartell™ 0046200
Product Code. 12509666
  • € 84.60 / Pack of 100
Estimated Shipment: 14-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Designed with serrations to hold hoses tightly and prevent slipping.
  • Made from polypropylene
  • Autoclavable
  • Chemical- and temperature-resistant
Compliance
Meet Food and Agriculture regulations
Specifications
Specifications
Meet with the Food and Drug regulations. 90/128/CEE 92/39/CEE e FDA 21 CFR 177.1520, CE and IVD
Polypropylene
100/Pk.
T-shape
Autoclavable
Suitable for foodstuff
6.3mm bore
Untapered T Tubing Connector
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
2 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberT23I03921 Certificate TypeDeclaration of Conformity Date01/08/2024 Catalog Number0046200
Lot NumberT23S00447 Certificate TypeDeclaration of Conformity Date06/07/2023 Catalog Number0046200
Special Offers
Special Offers
Product Content Correction
Product Title
Kartell™ Plastilab™ Untapered T-Shaped Tubing Connectors > T-shaped; Bore Size: 6.3mm; 100 Pack