Learn More
Steris™ Plain Top Closure Pouches
For use in porous load pre-vacuum autoclaves as well as gamma sterilizers. Steris Corporation™ Plain Top Closure Pouches include a double folded bottom seal for extra integrity.
€ 159.00 - € 213.00
Specifications
Autoclavable | Autoclavable |
---|---|
Certifications/Compliance | BS 6257 |
Closure Type | Plain |
Material | Medical Grade Kraft Paper (Bleached) |
Quantity | 250/Pk. |
Product Code | Brand | Dimensions (L x W x H) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W x H) | Price | Quantity & Availability | ||||||
15661964
|
Steris™
MPC |
300 x 75 x 530mm |
€ 159.00
Pack of 250 |
|
||||||
15681964
|
Steris™
T35PC |
380 x 125 x 610mm |
€ 213.00
Pack of 250 |
Estimated Shipment: 03-04-2025
Log in to see stock. |
||||||
Description
- Manufactured from high quality wet strength MG bleached kraft paper
- Double folded bottom seal for extra integrity
- Opening along width
- Conforms to BS 6257
Specifications
Autoclavable | |
Plain | |
250/Pk. |
BS 6257 | |
Medical Grade Kraft Paper (Bleached) |
Product Title
Steris™ Plain Top Closure Pouches