Learn More
Welch™ PIZA 101 Model Digital Vaccum Gauge
Product Code. 10319790 Shop All Welch Ilmvac Products
Click to view available options
Operating Temperature (Metric):
0°C to 60°C
Unit Size:
Each
Product Code. 10319790
Brand: Welch™ 600071
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Includes Sensor and 1m cable
Operating Temperature (Metric) 0°C to 60°C
Weight (Metric) 250 g
Pressure Range 1 to 1050mbar
Product Type Digital Vaccum Gauge