missing translation for 'onlineSavingsMsg'
Learn More

Hirschmann™ Pipetus™ Standard Pipet Filler

Provides the best prerequisites for working in potentially explosive environments, with toxic reagents or in clean room technology through the separation of the electric pump and hand-held unit. Hirschmann Instruments™ Pipetus™ Standard Pipet Filler enables highly accurate pipetting through fine dispensing valves with valve pins made of ceramic compound. A safety valve prevents the penetration of liquid into the interior of the unit.

Brand:  Hirschmann™ 9904000

 View more versions of this product

Product Code. 10203091

  • € 441.00 / Each
Estimated Shipment: 10-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Provides the best prerequisites for working in potentially explosive environments, with toxic reagents or in clean room technology through the separation of the electric pump and hand-held unit. Further sophisticated product details guarantee precision, reliability and a long service life. Highly accurate pipetting is enabled through fine dispensing valves with valve pins made of ceramic compound. A safety valve prevents the penetration of liquid into the interior of the unit. The standard PTFE diaphragm filter provided with 0.2μm pore size ensures reliable protection against overfilling. The sophisticated ergonomic shape of the pipetus™ standard aids non-fatiguing work.
  • Highly accurate pipetting enabled through fine dispensing valves with valve pins made of ceramic compound
  • The seat of all pipets is sealed absolutely tight with a silicone adapter
  • Extremely light hand unit with external pump
  • Hydrophobic PTFE diaphragm filter with 0.2 μm pore size
  • For measuring, volumetric and Pasteur pipettes, from 0.1 to 200mL
Specifications

Specifications

PTFE
0.2 μm
accurate pipetting
0.1 to 200mL
Euro
230 V
Standard
Pipette Filler
Measuring, volumetric, and Pasteur pipets
Filter on pump, pipet holder with safety valve and filter
Hand component: 130 g, pump, 290 g
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.