missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Terracycle™ Pipette Tip Boxes Zero Waste Box™

Click to view available options
Size:
Small
Medium
Large
Unit Size:
Each
Description
Use this Zero Waste Box to recycle:
- Empty pipette tip boxes
Do not send in:
- Pipette tips, tubes and rigid lab plastics that are contaminated with materials considered hazardous or dangerous
- Autoclaved material
- Hazardous waste (sharp, flammable, reactive, corrosive, ignitable, toxic, infectious or pathogenic) which presents a danger to the environment, or to people
- Batteries, pressurized canisters, broken glass and medical waste
Important:
- We do not collect any hazardous waste containing substances with toxic, reactive, corrosive, infectious, flammable, explosive properties or harmful to human health and the environment.
- Please make sure that you do not mix hazardous waste with non-hazardous waste.
Specifications
Specifications
| Capacity (Metric) | 25 kg |
| Capacity (English) | 55 lb. |
| Color | Beige, Green, Red, White |
| Dimensions (L x W x H) | 38 x 38 x 97 cm |
| Disposable | Single-use |
| Latex Free | Yes |
| Size | Large |
| Sterility | Non-sterile |
| Autoclavable | Not Autoclavable |
| Material | Cardboard, Plastic |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction