Learn More
Sartorius™ Pipette Calibration Kit
Shop All Sartorius Products
Click to view available options
For Use With (Equipment):
Cubis Balances
Unit Size:
Each
Description
- Used for models with 0.1 mg and 0.01 mg readability
- Consists of moisture trap and all required adapters
Spécification
Spécification
| Product Type | Calibration Kit |
| For Use With (Equipment) | Cubis Balances |