Learn More
Cole Parmer Pipette Bulb
Cole Parmer Pipette Bulb is sturdy, re-usable and easy-to-use.
Brand:  Cole Parmer 24820-60
Product Code. 11790299
  • € 40.80 / Pack of 3
Estimated Shipment: 03-02-2025
to see stock.
Explore more special offers
Description
Cole Parmer Pipette Bulb is sturdy, re-usable and easy-to-use.
  • Made from natural rubber and silicone
  • Can be used with Pasteur, serological, volumetric and other standard pipettes
  • Silicone seal grips the glass firmly and protects it
Specifications
Specifications
Rubber, Silicone
Non-sterile
25 mL