Learn More
Picropodophyllin, MedChemExpress
MedChemExpress Picropodophyllin (AXL1717) is a selective insulin-like growth factor-1 receptor (IGF-1R) inhibitor with an IC50 of 1 nM.
Brand: Medchem Express HY-15494
This item is not returnable.
View return policy
Description
Picropodophyllin (AXL1717) is a selective insulin-like growth factor-1 receptor (IGF-1R) inhibitor with an IC50 of 1 nM.Specifications
Picropodophyllin | |
White | |
C22H22O8 | |
AXL1717 Picropodophyllin PPP | |
O=C1OC[C@]2([H])[C@@H](O)C3=C(C=C4OCOC4=C3)[C@@H](C5=CC(OC)=C(OC)C(OC)=C5)[C@@]21[H] | |
414.41 | |
Research | |
Cancer-Kinase/protease |
477-47-4 | |
Research | |
50 mg | |
DMSO : 50 mg/mL (120.65 mM; Need ultrasonic) | |
414.41 | |
98.87% | |
Solid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Research purposes only